Learn More
Brady™ Self-laminating Vinyl MiniMark™ Wire and Cable Marker
Product Code. 11701825
Change view
Click to view available options
Width (Metric):
25.4 mm
38.1 mm
Height (Metric):
31.75 mm
63.50 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Height (Metric) unitSize
11701825 25.4 mm 31.75 mm Each
11721795 38.1 mm 63.50 mm Each
11791795 25.4 mm 63.50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11701825 Supplier Brady™ Supplier No. 710114
  • ONLINE EXCLUSIVE PRICE
    6875.00 NOK
    6409.90 NOK / Pack of 1
    Save 465.10 NOK
    6% Off
In Stock
Add to basket
Add to basket
This Self-laminating Vinyl MiniMark™ Wire and Cable Marker is made of conformable material designed to adhere to highly curved surfaces, wires and cables.
  • Features a white printable area with clear 'tail' that wraps around marker.
  • The clear tails serves as over-laminate to protect the text, providing superior resistance to abrasion, dirt, oils, solvents, water and excessive handling.
  • This material is UL recognised with its respective ribbon.
  • See the Recommended Ribbon Series for the appropriate ribbon to use.
Specifications
Certifications/Compliance ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognised
Description MiniMark Self-laminating Wire and Cable Markers
Adhesive Type Acrylic, Permanent
Color White, Transparent
Finish Matte
Format Label
Material Self-laminating Vinyl
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Data and telecommunications labelling, Electrical labelling, Laboratory labelling, Wire and cable labelling
For Use With (Equipment) MiniMark Printer
Min. Operating Temperature (Metric) -70°C
No. per Pack 2780 Labels/Box
Quantity 2780 Pk.
Size (Metric) 25.4 x 31.75 mm (W x H)
Temperature Range (English) -94°F to +158°F
Temperature Range (Metric) -70°C to +70°C
Tensile Strength Tensile Strength and Elongation 12 lb./in. (210 N/100 mm), 152%, 11 lb./in. (193 N/100 mm), 195%
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (L x W) 31.75 x 25.40 mm
Length (English) 1.25 in.
Length (Metric) 31.75 mm
Width (English) 1 in.
Width (Metric) 25.4 mm
Height (English) 1.25 in.
Height (English) Printable Area 0.5 in.
Height (Metric) 31.75 mm
Height (Metric) Printable Area 12.7 mm
Diameter (Metric) 6.10 to 3.40 mm
Alternate Part Number MNK-BPT-311-427