Learn More
Brady™ Self-laminating Vinyl Link Label
Click to view available options
Color:
Red, Transparent
White
White, Transparent
Width (Metric):
12.70 mm
25.4 mm
38.1 mm
Height (Metric):
25.4 mm
38.1 mm
Unit Size:
1 roll
Pack of 250
Description
- Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text
- Durable, permanent acrylic adhesive has outstanding water and oil resistance
- RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved
- Service temperature of -40°F to 158°F (-40°C to 70°C)
Specifications
Specifications
| Certifications/Compliance | RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC, UL Recognized |
| Description | TLS 2200/TLS PC Link Labels |
| Adhesive Type | Acrylic, Permanent |
| Color | White |
| Finish | Matte |
| Format | Self-laminating |
| Material | Vinyl |
| Print Technology | Thermal Transfer |
| Product Type | Link Label |
| Sterility | Non-sterile |
| Show More |