Learn More
Brady™ Self-laminating Polyester Labels for M710
Click to view available options
Dimensions (LxW):
15 x 40 mm
22.86 x 22.86 mm
25.4 x 25.4 mm
38.1 x 15.24 mm
41.28 x 15.24 mm
44.45 x 25.4 mm
45.72 x 9.53 mm
48.26 x 15.24 mm
66.68 x 15.24 mm
66.68 x 25.4 mm
Color:
White
White, Transparent
Shape:
Rectangle
Rectangle with Circle
Square
missing translation for 'unitSize'
Pack of 100
Pack of 150
Pack of 250
Pack of 500
Description
- Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
- Good print smudge resistance, solvent resistance and high/low temperature performance
Specifications
Specifications
| Certifications/Compliance | ASTM D 1000 |
| Adhesive Type | Permanent Acrylic |
| Coating | None |
| Color | White, Transparent |
| Finish | Matte |
| Format | Label |
| Material | Polyester |
| Printable Sides | Single-side Printable |
| Print Technology | Thermal Transfer |
| Product Type | Self-laminating Tag |
| Show More |