Learn More
Selenium, Black 99+, MilliporeSigma™
Product Code. 15835020
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15835020 1 kg Plastic bottle 1kg
15815020 50 g Plastic bottle 50g
15825020 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15835020 Supplier Supelco Supplier No. 1077141000
 Restricted Item
Estimated Shipment: 19-11-2026
Chemical Identifiers
CAS 7782-49-2
Molecular Formula Se
Molecular Weight (g/mol) 78.97
MDL Number MFCD00134090 MFCD00011224
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
Synonym selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
PubChem CID 6326970
ChEBI CHEBI:27568
IUPAC Name selenium
SMILES [Se]
Specifications
Physical Form Solid
Quantity 1 kg
Formula Weight 78.96
Odor Odorless
Packaging Plastic bottle
Appearance Dark gray solid
Chemical Name or Material Selenium