Learn More
Thermo Scientific™ Selenite Lactose Broth
Click to view available options
Quantity:
50/Pk.
Media Type:
Selenite Lactose Broth
Fill Volume:
8 mL
Unit Size:
Pack of 50
Description
Enrich for Salmonella species from fecal and food samples with Thermo Scientific™ Selenite Lactose Broth in convenient prepared media tubes.
Selenite Lactose Broth is used for enrichment culture of salmonellae from feces, food products, water and other materials.
Sodium biselenite acts as the selective agent, while lactose is added as a fermentable carbohydrate to prevent a rise in pH value during incubation because any increase in pH will reduce the selective activity of selenite.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Sodium biselenite acts as the selective agent, while lactose is added as a fermentable carbohydrate to prevent a rise in pH value during incubation because any increase in pH will reduce the selective activity of selenite.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | Light ivory, transparent |
| Content And Storage | 2°C to 12°C |
| Format | Tube |
| Manufacturing Origin | Germany |
| Media Type | Selenite Lactose Broth |
| Product Type | Microbiology Media |
| Fill Volume | 8 mL |
| Fill Weight | 8 ± 0.5mL |
| Packaging Type | 50 tubes in a carton box |
| Quantity | 50/Pk. |
For In Vitro Diagnostic Use.