Description
- SecurityGuard™ Cartridges for use with Reverse Phase HPLC columns
- Available in multiple internal diameter options
- All columns are 4 mm in length
Specifications
Specifications
| Quantity | 10 Pk. |
| Particle Size | Proprietary |
| Length (Metric) | 4 mm |
| Type | Guard Cartridge |
| Diameter (Metric) Inner | 3 mm |
| Stationary Phase | Gemini C18 |
| Technique | Reverse Phase HPLC |
| Product Line | SecurityGuard |