Learn More
(-)-Securinine, MedChemExpress
Product Code. 16449258
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449258 10 mg 10mg
16411678 5 mg 5mg
16403946 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16449258 Supplier Medchem Express Supplier No. HYN2079
MedChemExpress (-)-Securinine is plant-derived alkaloid and also a GABAA receptor antagonist.
(-)-Securinine is plant-derived alkaloid and also a GABAA receptor antagonist.
Specifications
Chemical Name or Material (-)-Securinine
CAS 5610-40-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C13H15NO2
Quantity 10 mg
Solubility Information DMSO : 2 mg/mL (9.21 mM; Need ultrasonic)
SMILES O=C(O1)C=C2[C@@]13[C@](CCCC4)([H])N4[C@](C3)([H])C=C2
Molecular Weight (g/mol) 217.26
Formula Weight 217.26
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death