Learn More
3M™ SecureFit™ X5500 Safety Helmet, Vented, CE
Product Code. 17385333
Change view
Click to view available options
Color:
Black
Blue
Green
Hi-vis Green
Orange
Red
Yellow
Unit Size:
Pack of 4
Product Code. Color unitSize
17385333 Black Pack of 4
17335333 Yellow Pack of 4
17345333 Blue Pack of 4
17355333 Green Pack of 4
17365333 Red Pack of 4
17375333 Orange Pack of 4
17395333 Hi-vis Green Pack of 4
7 options
Product Code. 17385333
Brand: 3M™ 7100175597
Only null left
Add to basket
Add to basket
3M™ SecureFit™ Safety Helmet with adjustable positioning and easy-to-adjust, 4-point ratchet suspension system that features 3M ™ Pressure Diffusion Technology. 3M™ UVicator™ Sensor indicates over-exposed to UV-radiation.
  • Secure, comfortable head protection against small falling objects and for general industrial use
  • Suspension system incorporates exclusive patented 3M™ Pressure Diffusion Technology
  • 4-point ratchet suspension allows wearer to customise height and fit with multiple levels of vertical adjustment
  • Vents allow airflow through the helmet
  • Optional vents allow airflow through the helmet
  • Inspired by climbing helmets with a brimless design to give you a clear upward view
  • 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV-radiation and it's time to replace it
  • 3M™ Scotchlite™ Reflective Material can be added to increase visibility under low light conditions
  • 3M™ Scotchlite™ Reflective Material can also be added to increase visibility under low light conditions
  • Approved for lateral deformation (LD)
  • Approved for lateral deformation
TRUSTED_SUSTAINABILITY
Specifications
Color Black
For Use With (Application) Construction, Forestry, Oil, and Gas
Material ABS
Product Type Safety Helmet
Suspension Type 4-point Ratchet
Ventilated Ventilated