Learn More
3M™ SecureFit™ X5500 Safety Helmet, Vented, CE
Description
- Secure, comfortable head protection against small falling objects and for general industrial use
- Suspension system incorporates exclusive patented 3M™ Pressure Diffusion Technology
- 4-point ratchet suspension allows wearer to customise height and fit with multiple levels of vertical adjustment
- Vents allow airflow through the helmet
- Optional vents allow airflow through the helmet
- Inspired by climbing helmets with a brimless design to give you a clear upward view
- 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV-radiation and it's time to replace it
- 3M™ Scotchlite™ Reflective Material can be added to increase visibility under low light conditions
- 3M™ Scotchlite™ Reflective Material can also be added to increase visibility under low light conditions
- Approved for lateral deformation (LD)
- Approved for lateral deformation
Specifications
Specifications
| Color | Orange |
| For Use With (Application) | Construction, Forestry, Oil, and Gas |
| Material | ABS |
| Product Type | Safety Helmet |
| Suspension Type | 4-point Ratchet |
| Ventilated | Ventilated |