Learn More
3M™ SecureFit™ 200 Safety Glasses
Description
- Economical lightweight (18 g) 3M™ SecureFit™ Safety Glasses range
- 3M's innovative SecureFit™ Pressure Diffusion Temple technology design features slotted ribs that provide comfortable pressure equalization.
- The temples flex and self-adjust to the size of wearer's head
- Scotchgard™ Anti-Fog Coating provides superior anti-fog and anti-scratch properties (available on selected models)
- Wraparound design improves the security of fit and prevents slipping
- Flat and flexible temples combine easily with earmuffs
- Easy to clean
- Foam gasket available to help limit eye exposure to debris
- Prescription insert available
Specifications
Specifications
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Tint | Gray |
| Product Type | Safety Glasses |
| Material | Plastic |
| Disposable | Reusable |
| Lens Material | Plastic |
| Frame Material | Plastic |
| UV Protection | None |
| Frame Type | Standard |