Learn More
3M™ Secure Click™ Particulate Filters
3M™ Secure Click™ Particulate Filter is our CE approved filter for environments containing certain particles.
1520.00 NOK - 1880.00 NOK
Specifications
Certifications/Compliance | CE Approved |
---|---|
Connection Type | Secure Click™ |
Product Type | Filter |
Quantity | 80/Cs. |
Product Code | Brand | Filter Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Type | Price | Quantity & Availability | |||||
16466390
|
3M™
7100197481 |
P2R |
1520.00 NOK
10 pairs |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
16426390
|
3M™
7100197284 |
P3R |
1880.00 NOK
10 pairs |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
Description
- New dual flow cartridges and filters allow for easier breathing
- Protects against particulates with optional nuisance level organic vapour/acid gas relief
- Quick, simple connection. Simply align the connections and push until you hear a click
- Use with 3M™ Secure Click™ Reusable Respirators 800 Series
- Swept-back design allows an enhanced field of view and comfort
Specifications
CE Approved | |
Secure Click™ | |
Filter | |
80/Cs. |