Learn More
3M™ Secure Click™ Half Mask Reusable Respirator
Product Code. 16486370 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
Includes:
Speaking Diaphragm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Includes Unit Size
16486370 Small Speaking Diaphragm Each
16446370 Medium Speaking Diaphragm Each
16476370 Large Speaking Diaphragm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16486370 Supplier 3M™ Supplier No. 7100172006
  • ONLINE EXCLUSIVE PRICE
    791.00 NOK
    532.62 NOK / Pack of 1
    Save 258.38 NOK
    32% Off
View all promotions
Estimated Shipment: 19-11-2026
Designed with smart and intuitive features, the Secure Click™ Half Mask Reusable Respirator is simple, comfortable and reliable. Available in three sizes: small, medium, and large, these respirators all come with an optional speaking diaphragm.
  • Greater breathability and comfort provided by the world's first quad flow cartridge system.
  • Push button seal check helps bolster confidence that the respirator is properly sealed to the face.
  • Speaking diaphragm is designed to help provide easier communication while you work.
  • Confidence that filters and cartridges are installed properly provided by all new Secure Click™ connection.
  • Low profile half mask design offers a wide field of view and compatibility with certain welding and grinding shields.
  • Over-moulded design for easy assembly/disassembly.
  • Advanced silicone material for increased comfort and greater durability.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance AS/NZS 1716:2012
Connection Type Secure Click™
Color Dark Gray
Material Silicone, Nylon
Product Type Half Facepiece Respirator
Size Small
Includes Speaking Diaphragm
Model HF-801 SD
Quantity 10/Cs.