Learn More
SEC63 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28665-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16876205
  • 4593.76 NOK / 150µL
Estimated Shipment: 27-05-2024
Description
The SEC63 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SEC63. This antibody recognizes human, mouse antigen. The SEC63 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
SEC63
Polyclonal
Unconjugated
150 μL
11231
IgG
Western Blot, Immunohistochemistry, Immunofluorescence
333 μg/mL
Rabbit
Primary
Human, Mouse