Learn More
SEC63 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28665-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The SEC63 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SEC63. This antibody recognizes human, mouse antigen. The SEC63 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
SEC63 | |
Polyclonal | |
Unconjugated | |
150 μL | |
11231 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
333 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |