missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SEC63 Polyclonal antibody specifically detects SEC63 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | SEC63 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 750 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | PRO2507, SEC63 homolog (S. cerevisiae), SEC63LERdj2, SEC63-like (S. cerevisiae), translocation protein SEC63 homolog |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 595-720 of human SEC63 (NP_009145.1).,, Sequence:, RDSDREQDEKQNKDDEAEWQELQQSIQRKERALLETKSKITHPVYSLYFPEEKQEWWWLYIADRKEQTLISMPYHVCTLKDTEEVELKFPAPGKPGNYQYTVFLRSDSYMGLDQIKPLKLEVHEAK |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?