missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC5/EXOC2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1C11G2, Proteintech
Shop All Proteintech ProductsDescription
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and the functions of the exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. This interaction has been shown to mediate filopodia formation in fibroblasts.
Specifications
Specifications
| Antigen | SEC5/EXOC2 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 1C11G2 |
| Concentration | 1.22 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | EXOC2 |
| Gene Accession No. | O54921, Q96KP1, Q9D4H1 |
| Gene Alias | EXOC2, exocyst complex component 2, Exocyst complex component Sec5, SEC5, SEC5L1, Sec5p |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?