Learn More
SEC14L1 Rabbit anti-Human, Polyclonal, Proteintech
Product Code. 16894105 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16894105 150 μL 150µL
16804115 20 μL 20µL
2 options
Product Code. 16894105
Brand: Proteintech 255411AP150UL
Rabbit Polyclonal Antibody
SEC14L1 may play a role in innate immunity by inhibiting the antiviral RIG-I signaling pathway. In this pathway, functions as a negative regulator of DDX58/RIG-I, the cytoplasmic sensor of viral nucleic acids. Prevents the interaction of DDX58 with MAVS/IPS1, an important step in signal propagation. May also regulate the SLC18A3 and SLC5A7 cholinergic transporters.
Specifications
Antigen SEC14L1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SEC14L1
Gene Accession No. Q92503
Gene Alias PRELID4A, SEC14 like 1 (S. cerevisiae), SEC14 like protein 1, SEC14L, SEC14L1
Gene Symbols SEC14L1
Host Species Rabbit
Immunogen SEC14L1 Fusion Protein Ag22233
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6397
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG