Learn More
Buerkle™ Sealing Sleeves for Buerkle™ Zone Samplers
Product Code. 11886413 Shop All Bürkle Products
Change view
Click to view available options
Diameter (Metric) Outer Top:
25 mm
50 mm
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Top Unit Size
11886413 50 mm 1 set
10770763 25 mm 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11886413 Supplier Buerkle™ Supplier No. 53170010
Estimated Shipment: 14-01-2027
Sealing sleeves made of PP, sterilisable, they seal individual chambers when filling, emptying or transporting. The sealing sleeves are easy to slide on and they automatically clip into position.
Sealing sleeves made of PP, sterilizable, which seal individual chambers when filling, emptying or transporting. The sealing sleeves are easy to slide on and automatically clip into position. Length 150 mm
TRUSTED_SUSTAINABILITY
Specifications
Material PVC
For Use With (Equipment) Zone Sampler
Diameter (Metric) Outer Top 50 mm
Length (Metric) 150 mm