missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDF2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14747-1-AP-150UL
This item is not returnable.
View return policy
Description
SDF2 Polyclonal antibody apecifically detetects SDF2 antigen in Human, Mouse Samples. It is validated for Western Blot.Specifications
| SDF2 | |
| Polyclonal | |
| Unconjugated | |
| BC000500 | |
| Rabbit | |
| Antigen affinity purification | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Western Blot | |
| 500 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| SDF2 | |
| SDF2 fusion protein Ag6480 | |
| 150 μL | |
| 6388 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction