Learn More
SDC1 Monoclonal Antibody (OTI7H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813775
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
SDC1 Monoclonal antibody specifically detects SDC1 in Human samples. It is validated for Flow CytometrySpecifications
| SDC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P18827 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 6382 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry | |
| OTI7H2 | |
| Unconjugated | |
| Sdc1 | |
| AA408134; AA409076; CD antigen 138; CD138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; HSPG; sCD138; SDC; Sdc1; soluble CD138; Sstn; syn-1; Synd; SYND1; Synd-1; SYNDECA; syndecan; syndecan 1; syndecan proteoglycan 1; Syndecan1; syndecan-1; synstatin | |
| Human recombinant protein fragment corresponding to amino acids 23-254 of human SDC1 (NP_001006947) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.