Learn More
Lenz Laborglasinstrumente™ Screwthread Tubes for Glassblowers
Product Code. 10693351
Change view
Click to view available options
Overall Length (Metric):
140 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Overall Length (Metric) Unit Size
10693351 140 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10693351 Supplier Lenz Laborglasinstrumente™ Supplier No. 01300032
  • ONLINE EXCLUSIVE PRICE
    720.00 NOK
    508.19 NOK / Pack of 10
    Save 211.81 NOK
    29% Off
Estimated Shipment: 27-01-2027
Manufactured from borosilicate glass. Lenz Laborglasinstrumente™ Screwthread Tubes for Glassblowers has a high resistance to thermal shock and chemical corrosion. The tube combined with plastic fittings produce a tight connection to a straight tube.
  • Acc. To DIN 12216 and in addition to standar
  • With DIN thread (GL)
  • Material: DURAN™
Specifications
Certifications/Compliance DIN 12216
Diameter (Metric) 28 mm
Material Borosilicate Glass (3.3)
Overall Length (Metric) 140 mm
Thickness (Metric) 2.0 mm
Packaging Quantity 10
Thread Size 32