Learn More
Thermo Scientific™ Screw Vial Convenience Kit
Product Code. p-10162037
Click to view available options
Volume (Metric):
2 mL
4 mL
8 mL
12 mL
16 mL
Color:
Amber
Clear
Closure Type:
Screw Cap, Open Top
Solid Top, Not Magnetic
Unit Size:
Pack of 100
Pack of 200
Product Code. 16595874
Brand: Thermo Scientific™ B780019A
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Storage vials range from 2mL to 40mL.
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
Order as individual sample storage vials and closures or as unassembled convenience kits.
Unassembled convenience kits saves time during sample preparation.
  • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
  • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Black
Description 8 mm Screw Amber Glass Vial Convenience Kit, Vial & Closure B7999-1A, B7815-9
Color Amber
Diameter (Metric) Outer 12 mm
Material Glass
Pre-slit No
Volume (Metric) 2 mL
Septum PTFE/Silicone liner
Includes 2mL amber glass vial, solid top PP screw cap
Closure Material Polypropylene
Closure Type Solid Top, Not Magnetic
Height (Metric) 32 mm
Quantity Pack of 100
Certifications/Compliance Specification Certified
Type Assembled, Unassembled Screw Vial Kit
Closure Size 8-425
Septa Size (Metric) 8-425 mm