Learn More
WTW™ Screw Connection
For use with OxiTop™ AD/SK/AD measuring systems.
Brand:  WTW™ 209500
Additional Details : Weight : 1.00000kg
Product Code. 10439332
  • 152.00 NOK / Each
Estimated Shipment: 13-06-2024
Description
  • Screwable connection for adapter
  • Aids in determination of soil respiration
Specifications
Specifications
45 thread
Screw Connection/Adapter
OxiTop AD/SK /OxiTop AD Adapter