Learn More
- Home
- Products
- Tubes
- Tube Caps and Closures
- Thermo Scientific™ Screw Cap Microcentrifuge Tube Caps
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
-
Actual product may vary.
Thermo Scientific™ Screw Cap Microcentrifuge Tube Caps
Product Code. p-6044000
Shop All MBP Molecular BioProducts Products
| Product Code. | Color | Product Type | Sterility | Fit for Purpose | Unit Size |
|---|---|---|---|---|---|
| 10695693 | Amber | Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16255570 | Amber | Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16265570 | Black | Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16275570 | Black | Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 10571303 | Blue | Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16285570 | Blue | Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 10357762 | Green | Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16295570 | Green | Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 13107673 | Natural | Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16265580 | Natural | Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 10042102 | Orange | Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16225580 | Orange | Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16235580 | Pink | Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16245580 | Pink | Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 10625893 | Red | Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16255580 | Red | Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16215610 | Violet | Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16225610 | Violet | Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16235610 | White | Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16245610 | White | Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 10002092 | Yellow | Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16255610 | Yellow | Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16205580 | Assorted (Black, Blue, Green, Natural, Orange, Pink, Red, Violet, White, Yellow) | Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16215580 | Assorted (Black, Blue, Green, Natural, Orange, Pink, Red, Violet, White, Yellow) | Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16285580 | Amber | Tether Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16295580 | Amber | Tether Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16215590 | Black | Tether Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16225590 | Black | Tether Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16205590 | Blue | Tether Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16235590 | Blue | Tether Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16245590 | Green | Tether Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16255590 | Green | Tether Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16275580 | Natural | Tether Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16245600 | Natural | Tether Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16285590 | Orange | Tether Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16295590 | Orange | Tether Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16205600 | Pink | Tether Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16215600 | Pink | Tether Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16225600 | Red | Tether Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16235600 | Red | Tether Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16255600 | Violet | Tether Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16265600 | Violet | Tether Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16275600 | White | Tether Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16285600 | White | Tether Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16295600 | Yellow | Tether Screw Cap with O-ring | Non-sterile | PCR Ready | Pack of 5000 |
| 16205610 | Yellow | Tether Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
| 16265590 | Assorted (Black, Blue, Green, Natural, Orange, Pink, Red, Violet, White, Yellow) | Tether Screw Cap with O-ring | - | PCR Ready | Pack of 5000 |
| 16275590 | Assorted (Black, Blue, Green, Natural, Orange, Pink, Red, Violet, White, Yellow) | Tether Screw Cap with O-ring | Sterile | Sterile and PCR Ready | Pack of 5000 |
Description
Prevent leakage under demanding conditions with Thermo Scientific™ Screw Cap Microcentrifuge Tube Caps feature O-Rings to form a secure seal. Optimized for secure, short-term storage in low temperatures, these non-autoclavable screw caps are available in a variety of colors and adapted to your scientific experiments.
Screw Cap Microcentrifuge Tube Caps Design
- Screw caps with O-rings guard against leakage by forming a positive seal against the rim of the tube
- Tethered screw caps remain attached to the sample tube but can rotate completely for convenience--the tethered screw cap is the solution when it is important to maintain the same cap with the same tube
- O-ring is manufactured from durable ethylene polypropylene dimonomere compound
Highest Quality
- Certified free of RNase, DNase, DNA and pyrogens
Color/Packaging Options
- Standard Screw Cap Color options are: Blue, Green, Orange, Red, Yellow and Mixed to aid with visual identification of samples
- Tethered screw cap with O-ring: Natural
Specifications
Specifications
| Product Type | Screw Cap with O-ring |
| Autoclavable | Not Autoclavable |
| Material | Polypropylene, Ethylene Propylene |
| Color | Amber |
| Packaging | 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag. |
| Clean Claims | Non-Sterile Free of: Rnase/Dnase, Human and Mouse DNA, Pyrogen & Endotoxin, ATP, PCR Inhibitor |
| UV Resistance | (UV) Ultra violet protection avg. 1.22% transmittance between 220 and 500nm |
| Binding Property | Low Nucleic Acid and Protein Binding & Low Enzyme Inhibition |
| Max. Torque | Manual/Hand Torque: Standard: 0.125 lbsf.in to 2.5 lbsf.in; Metric: 1.412 Ncm to 28.24 Ncm. Automated/Robotic Torque Standard: 1.312 lbsf.in to 2.5 lbsf.in; Metric: 14.83 Ncm to 28.24 Ncm |
| Temperature Range (Metric) | Cold Storage: -80° to +95°C. Liquid Nitrogen: Short term Vapor Phase & Quick Flash Freeze acceptable, not recommmended for LN2 submersion storage |
| Sterility | Non-sterile |
| Max. RCF | 25,000 x g |
| Fit for Purpose | PCR Ready |
| Show More Show Less |
Screw cap micro tubes and caps are not recommended for autoclave. Sterile options available.
Not intended to submerge and store in liquid phase LN2.