Learn More
Gosselin™ Screw Cap Dippers with Removable Handle
GREENER_CHOICE
Product Code. p-7015022
Change view
Click to view available options
Capacity (Metric):
40 mL
125 mL
180 mL
Unit Size:
Pack of 100
Pack of 250
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15488794 40 mL Pack of 250
15498794 125 mL Pack of 100
15408804 180 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15488794 Supplier Gosselin™ Supplier No. TAT30C01
Add to basket
Add to basket
Dip with confidence. Gosselin™ Screw Cap Dippers with Removable Handle prevent spillage with screw caps and detachable shaft.
  • Polypropylene blue containers
  • Removable handle
  • Shock resistant
  • Packaged individually in tamper evident single zip bags
  • Traceability information on each bag
  • Sterile SAL 10-3
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Capacity (Metric) 40 mL
Sterility Sterile
Color Blue
Diameter (Metric) Outer 43 mm
Height (Metric) 80 mm
Length (Metric) Handle 290 mm including container