Learn More
Gosselin™ Screw Cap Dippers with Removable Handle
Product Code. 15498794
Change view
Click to view available options
Capacity (Metric):
40 mL
125 mL
180 mL
Unit Size:
Pack of 100
Pack of 250
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15498794 125 mL Pack of 100
15488794 40 mL Pack of 250
15408804 180 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15498794 Supplier Gosselin™ Supplier No. TAT52C01
  • ONLINE EXCLUSIVE PRICE
    2935.00 NOK
    1544.64 NOK / Pack of 100
    Save 1390.36 NOK
    47% Off
Estimated Shipment: 29-01-2027
Dip with confidence. Gosselin™ Screw Cap Dippers with Removable Handle prevent spillage with screw caps and detachable shaft.
  • Polypropylene blue containers
  • Removable handle
  • Shock resistant
  • Packaged individually in tamper evident single zip bags
  • Traceability information on each bag
  • Sterile SAL 10-3
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Capacity (Metric) 125 mL
Sterility Sterile
Color Blue
Diameter (Metric) Outer 52 mm
Height (Metric) 74 mm
Length (Metric) Handle 275 mm including container