Learn More
Ohaus™ Screw Cap, 100 mL Bucket, Frontier™ R-S4x100/5MIS Swing Out Rotors
Product Code. 15886692 Shop All Ohaus Products
Click to view available options
:
For use with 100 mL Bucket, Frontier™ R-S4x100/5MIS Swing Out Rotors, O-rings
Product Code. 15886692
Brand: Ohaus™ 30314862
Securely seals centrifuge bucket, protecting centrifuge from potential spills.
  • Suitable for use with 100 mL buckets for R-S4x100/5MIS Swing Out Rotors
  • Requires 100 mL bucket cap O-rings
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 100 mL Bucket, Frontier, R-S4x100/5MIS Swing Out Rotors, O-rings
Description Screw Cap, 100ml Bucket, Sealable, 2/pk