Learn More
Fisherbrand™ Screen
Artikelnummer. 12358546
Change view
Click to view available options
For Use With (Equipment):
Fisherbrand GD-5040 Gel Dryer System
Packungsgröße:
Each
Artikelnummer. For Use With (Equipment) unitSize
12358546 Fisherbrand GD-5040 Gel Dryer System Each
10589391 Fisherbrand GD-5040 Gel Dryer System Each
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 12358546
Marke: Fisherbrand™ GDSSS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Accessory for use with vacuum gel dryers
  • Acts in conjunction with cast-aluminum base
  • Serve as porous gel support
  • Uniformly distributesthe heat and vacuum
  • Prevent gel cracking during drying
  • Pore size of stainless steel sheet: 1mm
Spezifikation
For Use With (Equipment) Fisherbrand GD-5040 Gel Dryer System
Type Screen