Learn More
BOLA Scrapers
Product Code. p-200110013
Change view
Click to view available options
Width (Metric):
50 mm
90 mm
120 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) unitSize
10309381 50 mm Each
10610372 90 mm Each
10013551 120 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10309381 Supplier BOLA Supplier No. H91602
Scrapers PTFE, solid
  • Scrapers made of PTFE with tapered end.
  • Ideal handling due to big handle and wide blade.
  • Universal chemical resistance.
Specifications
Sterility Non-sterile
Autoclavable Autoclavable
Length (Metric) 160 mm
Width (Metric) 50 mm