Learn More
Kimberly-Clark™ Scott™ Extra Couch Cover (51W) 6003, 6 rolls x 130
Description
- 6 rolls x 130 white, 2 ply sheets (810 total)
- Ideal for protecting couches and maintaining high levels of hygiene in the healthcare industry
- Comes in a lightweight and portable roll
- Soft, absorbent, tear-resistant and durable
- Quality protection
Specifications
Specifications
| Product Type | Couch Cover |
| Color | White |
| Material | Paper |
| Sterility | Non-sterile |
| Dimensions (L x W) | 50 x 37 cm |
| Format | Sheets |