Learn More
3M™ Scotch™ Performance Box Sealing Tape
Click to view available options
Dimensions (LxW):
660 m x 55 mm
990 m x 50 mm
Format:
Roll
Tape
Unit Size:
6 rolls
Description
- Reseals and packages heavy weight goods
- High initial tack and shear adhesion to fiberboard
- Use for single strip closure of fiberboard boxes
- Performs well at temperatures typically encountered by packaged products in shipping and storage environment
- Quality tape for closing boxes
- Resistant to fissures due to brutal manipulation and absorbs shocks
Specifications
Specifications
| Adhesive Type | Hot-melt |
| Color | Transparent |
| Format | Roll |
| Product Type | Box Sealing Tape |
| Model | 3739 |
| Dimensions (LxW) | 990 m x 50 mm |
| Length (English) | 1083 yd. |
| Length (Metric) | 990 m |
| Width (English) | 2 in. |
| Width (Metric) | 50 mm |