missing translation for 'onlineSavingsMsg'
Learn More

Scopoletin, 95%

Product Code. 10570404 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
50mg
This item is not returnable. View return policy

Product Code. 10570404

Brand: Thermo Scientific Acros 302902500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92-61-5 | C10H8O4 | 192.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-61-5
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00006872
InChI Key RODXRVNMMDRFIK-UHFFFAOYSA-N
Synonym scopoletin, gelseminic acid, 6-methylesculetin, 7-hydroxy-6-methoxy-2h-chromen-2-one, chrysatropic acid, scopoletine, murrayetin, scopoletol, 7-hydroxy-6-methoxycoumarin, 6-o-methylesculetin
PubChem CID 5280460
ChEBI CHEBI:17488
IUPAC Name 7-hydroxy-6-methoxychromen-2-one
SMILES COC1=C(C=C2C(=C1)C=CC(=O)O2)O

Specifications

Melting Point 200.0°C to 207.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 8545
Formula Weight 192.17
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Scopoletin, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.