Learn More
SCNM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16834694 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16834694 20 μL 20µL
16824694 150 μL 150µL
2 options
Product Code. 16834694
Brand: Proteintech 154901AP20UL
Rabbit Polyclonal Antibody
SCNM1 is a zinc finger protein and putative splicing factor. In mice, Scnm1 modifies phenotypic expression of Scn8a mutations.
Specifications
Antigen SCNM1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SCNM1
Gene Accession No. Q8K136, Q9BWG6
Gene Alias SCNM1, sodium channel modifier 1
Gene Symbols SCNM1
Host Species Rabbit
Immunogen SCNM1 Fusion Protein Ag7854
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310662, 69269, 79005
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG