missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCLY Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10667-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody detects the N-terminal of the protein.
Selenocysteine lyase (SCLY; EC 4.4.1.16) catalyzes the pyridoxal 5-prime phosphate-dependent conversion of L-selenocysteine to L-alanine and elemental selenium (Mihara et al., 2000).Specifications
| SCLY | |
| Polyclonal | |
| Unconjugated | |
| SCLY | |
| hSCL, SCL, SCLY, selenocysteine lyase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 363285, 50880, 51540 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q68FT9, Q96I15, Q9JLI6 | |
| Scly | |
| SCLY Fusion Protein Ag0972 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction