Learn More
Eppendorf™ Scientific Shaker Flask Clamps
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers. Eppendorf™ Scientific Shaker Flask Clamps are of single piece stainless-steel construction for added strength and safety.
212.00 NOK - 1200.00 NOK
Specifications
Product Type | Clamp |
---|---|
Material | Stainless Steel |
For Use With (Equipment) | Innova I-Series and Excella Shakers |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
11326044
|
Eppendorf™
ACE-1000S |
1000 mL Erlenmeyer Flask |
560.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11306044
|
Eppendorf™
M1190-9002 |
250 mL Erlenmeyer Flask |
357.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11316044
|
Eppendorf™
M1190-9003 |
500 mL Erlenmeyer Flask |
420.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11386034
|
Eppendorf™
M1190-9000 |
50 mL Erlenmeyer Flask |
242.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
10487651
|
Eppendorf™
M1190-9004 |
25 mL Erlenmeyer Flask |
231.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11396034
|
Eppendorf™
M1190-9001 |
125 mL Erlenmeyer Flask |
252.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11336044
|
Eppendorf™
ACE-2000S |
2000 mL Erlenmeyer Flask |
875.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11389814
|
Eppendorf™
ACFE-2800S |
2800 mL Fernbach Flask |
1060.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
10706164
|
Eppendorf™
ACE-10S |
10 mL Erlenmeyer Flask |
212.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11379814
|
Eppendorf™
ACE-6000S |
6000 mL Erlenmeyer Flask |
1200.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11359814
|
Eppendorf™
ACE-4000S |
4000 mL Erlenmeyer Flask |
1000.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
Description
- Stainless-steel construction
- Includes mounting screws
- Clamps 2mL and above, with an extra girdle for added security during agitating flasks at high speeds
- Complete with required screws, and are fitted with coiled wire springs for added stability at elevated speeds
Specifications
Clamp | |
Stainless Steel | |
Innova I-Series and Excella Shakers |