Learn More
Eppendorf™ Lab Shaker Clamps
Product Code. 11386034
Change view
Click to view available options
Holds:
1 L Erlenmeyer Flasks
1 L Media Bottles
10 mL Erlenmeyer Flasks
125 mL Erlenmeyer Flasks
2 L Narrow Neck Erlenmeyer, Thomson or Corning Flasks
2 L Wide Neck Erlenmeyer Flasks
2.8 L Fernbach Flasks
25 mL Erlenmeyer Flasks
250 mL Erlenmeyer Flasks
3 L Erlenmeyer Flasks
4 L Erlenmeyer Flasks
5 L Erlenmeyer Flasks
50 mL Erlenmeyer Flasks
500 mL Erlenmeyer Flasks
500 mL Media Bottles, 450 or 600 mL TPP Bioreactors
6 L Erlenmeyer Flasks
Dimensions (L x W):
23 x 22 mm
31 x 26 mm
51 x 26 mm
68 x 37 mm
91 x 47 mm
102 x 48 mm
130 x 64 mm
158 x 72 mm
203 x 73 mm
210 x 99 mm
92 x 108 mm
108 x 150 mm
165 x 112 mm
189 x 112 mm
222 x 145 mm
244 x 100 mm
Unit Size:
Each
Product Code. Holds Dimensions (L x W) unitSize
11386034 50 mL Erlenmeyer Flasks 51 x 26 mm Each
10706164 10 mL Erlenmeyer Flasks 31 x 26 mm Each
10487651 25 mL Erlenmeyer Flasks 23 x 22 mm Each
11396034 125 mL Erlenmeyer Flasks 68 x 37 mm Each
11306044 250 mL Erlenmeyer Flasks 91 x 47 mm Each
11316044 500 mL Erlenmeyer Flasks 102 x 48 mm Each
11399814 500 mL Media Bottles, 450 or 600 mL TPP Bioreactors 92 x 108 mm Each
11326044 1 L Erlenmeyer Flasks 130 x 64 mm Each
11309824 1 L Media Bottles 108 x 150 mm Each
11698613 2 L Narrow Neck Erlenmeyer, Thomson or Corning Flasks 165 x 112 mm Each
11336044 2 L Wide Neck Erlenmeyer Flasks 158 x 72 mm Each
11389814 2.8 L Fernbach Flasks 203 x 73 mm Each
11346044 3 L Erlenmeyer Flasks 189 x 112 mm Each
11359814 4 L Erlenmeyer Flasks 210 x 99 mm Each
11369814 5 L Erlenmeyer Flasks 222 x 145 mm Each
11379814 6 L Erlenmeyer Flasks 244 x 100 mm Each
16 options
Product Code. 11386034
Brand: Eppendorf™ M11909000
Only null left
Add to basket
Add to basket
Unlock the potential of your experiments with Eppendorf's versatile range of shaker clamps. Securely fasten Erlenmeyer flasks, Thomson Ultra Yield™, Optimum Growth™, Tunair™, Corning™, and more to your universal platforms. Find the ideal clamp for every flask type.
  • Stainless-steel construction (not spot welded or riveted) for added strength and safety.
  • Clamps 2 L and above come with an extra girdle for added security when agitating flasks at high speeds.
  • Comes complete with required screws, and are fitted with coiled wire springs for added stability at elevated speeds.
TRUSTED_SUSTAINABILITY
Specifications
Holds 50 mL Erlenmeyer Flasks
Product Type Clamp
Material Stainless Steel
Dimensions (L x W) 51 x 26 mm
For Use With (Equipment) Innova I-Series and Excella Shakers