missing translation for 'onlineSavingsMsg'
Learn More

SCH772984, MedChemExpress

Product Code. p-200098504
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16478108

Brand: Medchem Express HY50846

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SCH772984 is a highly selective and ATP-competitive ERK inhibitor, with IC50s of 4 and 1 nM for ERK1 and ERK2, respectively. SCH772984 has antitumor activity in MAPK inhibitor-naïve and MAPK inhibitor-resistant cells containing BRAF or RAS mutations.

SCH772984 is a highly selective and ATP-competitive ERK inhibitor, with IC50s of 4 and 1 nM for ERK1 and ERK2, respectively. SCH772984 has antitumor activity in MAPK inhibitor-naïve and MAPK inhibitor-resistant cells containing BRAF or RAS mutations.

Specifications

Chemical Name or Material SCH772984
CAS 942183-80-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C33H33N9O2
Quantity 10 mg
Solubility Information DMSO : 48 mg/mL (81.68 mM; Need ultrasonic)
SMILES O=C([C@H](CC1)CN1CC(N(CC2)CCN2C(C=C3)=CC=C3C4=NC=CC=N4)=O)NC5=CC6=C(C=C5)NN=C6C7=CC=NC=C7
Molecular Weight (g/mol) 587.67
Formula Weight 587.67
Percent Purity 98.69%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.