missing translation for 'onlineSavingsMsg'
Learn More

SCH 546738, MedChemExpress

Product Code. p-200098681
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475557

Brand: Medchem Express HY10017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SCH 546738 is a potent, orally active and non-competitive CXCR3 antagonist, the affinity constant (Ki) of SCH 546738 binding to human CXCR3 receptor is determined to be 0.4 nM in multiple experiments.

SCH 546738 is a potent, orally active and non-competitive CXCR3 antagonist, the affinity constant (Ki) of SCH 546738 binding to human CXCR3 receptor is determined to be 0.4 nM in multiple experiments.

Specifications

Chemical Name or Material SCH 546738
CAS 906805-42-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H31Cl2N7O
Quantity 5 mg
Solubility Information DMSO : 4.5 mg/mL (9.14 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N)C1=NC(Cl)=C(N2CCN(C3CCN(CC4=CC=C(Cl)C=C4)CC3)[C@@H](CC)C2)N=C1N
Molecular Weight (g/mol) 492.44
Formula Weight 492.44
Percent Purity 98.18%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.