Learn More
SCH 546738, MedChemExpress
Product Code. 16475557
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16475557
Brand: Medchem Express HY10017
MedChemExpress SCH 546738 is a potent, orally active and non-competitive CXCR3 antagonist, the affinity constant (Ki) of SCH 546738 binding to human CXCR3 receptor is determined to be 0.4 nM in multiple experiments.
SCH 546738 is a potent, orally active and non-competitive CXCR3 antagonist, the affinity constant (Ki) of SCH 546738 binding to human CXCR3 receptor is determined to be 0.4 nM in multiple experiments.
Specifications
Chemical Name or Material SCH 546738
CAS 906805-42-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H31Cl2N7O
Quantity 5 mg
Solubility Information DMSO : 4.5 mg/mL (9.14 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N)C1=NC(Cl)=C(N2CCN(C3CCN(CC4=CC=C(Cl)C=C4)CC3)[C@@H](CC)C2)N=C1N
Molecular Weight (g/mol) 492.44
Formula Weight 492.44
Percent Purity 98.18%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.