Learn More
SCH-1473759 hydrochloride, MedChemExpress
Product Code. 16490986
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490986 200 mg 200mg
16486707 5 mg 5mg
16463998 10 mg 10mg
16440116 2 mg 2mg
16410357 50 mg 50mg
16405006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16490986 Supplier Medchem Express Supplier No. HY10483
MedChemExpress SCH-1473759 hydrochloride is an aurora inhibitor with IC50s of 4 and 13 nM for aurora A and B, respectively.
SCH-1473759 hydrochloride is an aurora inhibitor with IC50s of 4 and 13 nM for aurora A and B, respectively.
Specifications
Chemical Name or Material SCH-1473759 hydrochloride
Physical Form Solid
CAS 1094067-13-6
Color Off-White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C20H27ClN8OS
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 70 mg/mL (151.19 mM; Need ultrasonic)
SMILES [H]Cl.CC1=CN2C(C(NC3=CC(CN(C(C)(CO)C)CC)=NS3)=N1)=NC=C2C4=CNN=C4
Molecular Weight (g/mol) 463.0
Formula Weight 463.0
Percent Purity 98.04%
Grade Research