missing translation for 'onlineSavingsMsg'
Learn More

SCH-1473759 hydrochloride, MedChemExpress

Product Code. 16463998
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
2mg
50mg
5mg
Product Code. Quantity unitSize
16463998 10 mg 10mg
16490986 200 mg 200mg
16486707 5 mg 5mg
16440116 2 mg 2mg
16410357 50 mg 50mg
16405006 100 mg 100mg
6 options
This item is not returnable. View return policy

Product Code. 16463998

Brand: Medchem Express HY10483

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SCH-1473759 hydrochloride is an aurora inhibitor with IC50s of 4 and 13 nM for aurora A and B, respectively.

SCH-1473759 hydrochloride is an aurora inhibitor with IC50s of 4 and 13 nM for aurora A and B, respectively.

Specifications

Chemical Name or Material SCH-1473759 hydrochloride
Physical Form Solid
CAS 1094067-13-6
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H27ClN8OS
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 70 mg/mL (151.19 mM; Need ultrasonic)
SMILES [H]Cl.CC1=CN2C(C(NC3=CC(CN(C(C)(CO)C)CC)=NS3)=N1)=NC=C2C4=CNN=C4
Molecular Weight (g/mol) 463.0
Formula Weight 463.0
Percent Purity 98.04%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.