missing translation for 'onlineSavingsMsg'
Learn More

SCGN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16843834 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16843834 20 μL 20µL
16833834 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16843834

Brand: Proteintech 149191AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The encoded protein is a secreted calcium-binding protein which is found in the cytoplasm. It is related to calbindin D-28K and calretinin. This protein is thought to be involved in KCL-stimulated calcium flux and cell proliferation.

Specifications

Antigen SCGN
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SCGN
Gene Accession No. O76038, Q6R556, Q91WD9
Gene Alias CALBL, SCGN, SECRET, Secretagogin, SEGN, setagin
Gene Symbols SCGN
Host Species Rabbit
Immunogen SCGN Fusion Protein Ag6736
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10590, 214189, 306942
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.