missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCGN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14919-1-AP-150UL
This item is not returnable.
View return policy
Description
The encoded protein is a secreted calcium-binding protein which is found in the cytoplasm. It is related to calbindin D-28K and calretinin. This protein is thought to be involved in KCL-stimulated calcium flux and cell proliferation.Specifications
| SCGN | |
| Polyclonal | |
| Unconjugated | |
| SCGN | |
| CALBL, SCGN, SECRET, Secretagogin, SEGN, setagin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10590, 214189, 306942 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O76038, Q6R556, Q91WD9 | |
| SCGN | |
| SCGN Fusion Protein Ag6736 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction