missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCGBL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21142-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
SCGBL | |
Polyclonal | |
Unconjugated | |
SCGB2B2 | |
SCGBL, secretoglobin like, Secretoglobin like protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
284402 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4G0G5 | |
SCGB2B2 | |
SCGBL Fusion Protein Ag15610 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |