Learn More
Trajan Scientific and Medical™ SCeramic Capillary Tube Cutter
Product Code. 15238843
Change view
Click to view available options
For Use With (Equipment):
Fused Silica Tubing
Unit Size:
Pack of 3
Product Code. For Use With (Equipment) unitSize
15238843 Fused Silica Tubing Pack of 3
1 options
Product Code. 15238843
Brand: Trajan Scientific and Medical™ 0625011
Only null left
Add to basket
Add to basket
Ceramic Capillary Tubing Cutter. Pack of 3 wafers.
  • Burr-free cutting of fused silica tubing
  • Reduces likelihood of blockages
  • Clean entry for on-column injection
Specifications
Type Capillary Ceramic Tube Cutter
Quantity 3/Pk.
For Use With (Equipment) Fused Silica Tubing