missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SCEL (Human) Recombinant Protein
Human SCEL full-length ORF ( AAH47536.1, 1 a.a. - 668 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00008796-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSNVTLRKMSPTGNEMKSTTQGTTRKQQDFHEVNKRRTFLQDNSWIKKRPEEEKDENYGRVVLNRHNSHDALDRKVNERDVPKATISRYSSDDTLDRISDRNDAAKTYKANTLDNQLTNRSMSVFRSLEVTKLQPGGSLNANTSNTIASTSATTPVKKKRQSWFPPPPPGYNASSSTGTRRREPGVHPPIPPKPSSPVSSTNQLRQDNRQIHPPKPGVYTETNRSAERNISEELDNLIKMNKSLNRNQGLDSLFRANPKVEEREKRAKSLESLIYMSTRTDKDGKGIQSLGSPIKVNQRTDKNEKGRQNLESVAKVNARMNKTSRRSEDLDNATEVNPKGHENTTGKKDLDGLIKVDPETNKNITRGQSLDNLIKVTPEVKRSNQGSKDLNNFIKVYPGTEKSTEGGQSLDSLIKVTPERNRTNQGNQDLENLIKVIPSANKSSEQGLDEHINVSPKAVKNTDGKQDLDKLIKVNPEIFTNNQRNQDLANLIKVNPAVIRNNQSQDLDNLIKVKPSALRNTNRDQNLENLIEVNSHVSENKNGSSNTGAKQAGPQDTVVYTRTYVENSKSPKDGYQENISGKYIQTVYSTSDRSVIERDMCTYCRKPLGVETKMILDELQICCHSTCFKCEICKQPLENLQAGDSIWIYRQTIHCEPCYSKIMAKWIP
Specifications
AAH47536.1 | |
sciellin | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SCEL | |
GST |
8796 | |
Wheat germ expression system | |
10 μg | |
FLJ21667, MGC22531 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |