Learn More
Buerkle™ Scrapers for Foodstuffs
Product Code. p-7230558 Shop All Bürkle Products
Change view
Click to view available options
Length (Metric):
205 mm
250 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10260512 205 mm Each
10440401 250 mm Each
2 options
Product Code. 10260512
Brand: Buerkle™ 83004011
Only null left
Add to basket
Add to basket
Scrapers with easy-to-clean, smooth and pore-free surface ideal for the food sector.
  • For foodstuffs (FDA and EU Nr. 10/2011)
  • Polypropylene has a high resistance to chemicals and temperatures
  • Can be also used for boiling products
TRUSTED_SUSTAINABILITY
Specifications
Product Type Scraper
Handle Type Fixed
Autoclavable Autoclavable
Certifications/Compliance FDA and (EU) Nr. 10/2011
Color White
For Use With (Application) Suitable for Use with Foodstuffs
Length (Metric) 205 mm
Material Polypropylene
Style One-piece
Width (Metric) 75 mm