Learn More
Interscience™ Scan 4000
Product Code. 15928015
Change view
Click to view available options
Display Type:
None
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Type Unit Size
15928015 None Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15928015 Supplier Interscience™ Supplier No. 438000
  • ONLINE EXCLUSIVE PRICE
    360510.00 NOK
    351521.13 NOK / Pack of 1
    Save 8988.87 NOK
    2% Off
Estimated Shipment: 02-12-2026
Scan 4000 is an automatic HD color colony counter that adapts to all sizes of Petri dishes and any culture media. Its indirect lighting system gives ideal user comfort, high accuracy and reproducibility.
  • Allows reading on Petri dishes of up to 150 mm diameter, and on square Petri dishes of 120 mm
  • Colony counter with more flexibility in your work
  • Also reads inhibition zones on round or square Petri dishes
Specifications
Product Type Colony Counter
Display Type None
Dimensions (L x W x H) 47 x 47 x 64 cm
For Use With (Application) Colony Counting, Inhibition Zone Reading
For Use With (Equipment) DataLink Pro