missing translation for 'onlineSavingsMsg'
Learn More

SC75741, MedChemExpress

Product Code. p-200100785
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16470357

Brand: Medchem Express HY10496

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SC75741 is a broad and efficient NF-κB inhibitor with an IC50 of 200 nM for p65. SC75741 blocks influenza viruses (IV) replication. SC75741 impairs DNA binding of the NF-κB subunit p65, resulting in reduced expression of cytokines, chemokines, and pro-apoptotic factors. SC75741 subsequently inhibits caspase activation and blocks caspase-mediated nuclear export of viral ribonucleoproteins.

SC75741 is a broad and efficient NF-κB inhibitor with an IC50 of 200 nM for p65. SC75741 blocks influenza viruses (IV) replication. SC75741 impairs DNA binding of the NF-κB subunit p65, resulting in reduced expression of cytokines, chemokines, and pro-apoptotic factors. SC75741 subsequently inhibits caspase activation and blocks caspase-mediated nuclear export of viral ribonucleoproteins.

Specifications

Chemical Name or Material SC75741
CAS 913822-46-5
Color Brown
Purity Grade Notes Research
Molecular Formula C29H23N7O2S2
Quantity 50 mg
Solubility Information DMSO : 83.33 mg/mL (147.31 mM; Need ultrasonic)
SMILES O=C(NC1=NC(C=CC(C(C2=CC=CC=C2)=O)=C3)=C3N1)C4=CSC(C(CC5)CCN5C6=NC=NC7=C6SC=C7)=N4
Molecular Weight (g/mol) 565.67
Formula Weight 565.67
Percent Purity 98.02%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.