missing translation for 'onlineSavingsMsg'
Learn More

SC144 hydrochloride, MedChemExpress

Product Code. p-200101354
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476836

Brand: Medchem Express HY15614A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SC144 hydrochloride is a first-in-class, orally active gp130 (IL6-beta) inhibitor.

SC144 hydrochloride binds gp130, induces gp130 phosphorylation (S782) and deglycosylation, abrogates Stat3 phosphorylation and nuclear translocation, and further inhibits the expression of downstream target genes. SC144 hydrochloride shows potent inhibition of gp130 ligand-triggered signaling. SC144 hydrochloride induces apoptosis in human ovarian cancer cells.

Specifications

Chemical Name or Material SC144 hydrochloride
CAS 917497-70-2
Color Gray
Purity Grade Notes Research
Molecular Formula C16H12ClFN6O
Quantity 100 mg
Solubility Information DMSO : 10 mg/mL (27.87 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(C1=NC=CN=C1)NNC2=NC3=C(N4C2=CC=C4)C=CC(F)=C3.[H]Cl
Molecular Weight (g/mol) 358.76
Formula Weight 358.76
Percent Purity 98.89%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.